Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva SOURCE™ Tricorn™ Columns for IEX

Catalog No. 45000311 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Source 15Q prepacked column
Source 15S prepacked column
Ligand Type:
Quaternary Ammonium
Sulphonate group
pH Range:
2 to 12
2 to 13
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Ligand Type pH Range
45-000-311 Source 15Q prepacked column Quaternary Ammonium 2 to 12
45-000-312 Source 15S prepacked column Sulphonate group 2 to 13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-311 Supplier Cytiva Supplier No. 17518101
Add to Cart
Add to Cart

Includes

Tricorn columns have Valco fittings for AKTAdesign™ systems and are supplied with M6 connectors for FPLC Systems, instruction book, and two stop plugs attached to the column.

Source 15Q 4.6/100 PE is a strong anion exchanger prepacked in a Tricorn column for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.

  • SOURCE 15Q media is highly suitable for fast, high-resolution purifications and easy scale-up
  • Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics, sample loading of up to 25 mg of protein/ml, and improved stability in organic solvents and pH extremes
  • Improved capacity compared to MonoBeads but with a slightly lower resolution.

Recommended Storage

4 to 30°C, 20% Ethanol

TRUSTED_SUSTAINABILITY

Specifications

Buffer All common ion exchange Buffers
Product Type Source 15Q prepacked column
Bed Volume 1.7mL
Diameter (English) Inner 0.18 in.
Diameter (Metric) Inner 4.6mm
Quantity 1 x 1.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <5 mL/min.
Material PEEK
Ligand Type Quaternary Ammonium
Max. Pressure 40 bar
pH Range 2 to 12
Particle Size 15μm
Support Type Polystyrene/Divinylbenzene
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) High performance ion exchange chromato graphy of Proteins, Peptides, Polynucleotides and other biomolecules
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.