Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Cell Staining Buffer
SDP

Catalog No. ob022501m Shop All Southern Biotechnology Associates Products
Click to view available options
Quantity:
100 mL
250 mL
500 mL

For dilution and application of fluorescent reagents as well as for suspension, washing and storage of cells destined for flow cytometric analysis

Designed to maximize fluorescence signal intensities generated by pH-sensitive fluorochromes. In addition, this buffer contains a metabolic inhibitor sodium azide (NaN3) and animal serum proteins to help minimize non-specific binding.

Applications:

Flow cytometry, Direct immunofluorescence staining, Indirect immunofluorescence staining, Intracellular staining, Immunocytochemistry

Specifications

Product Type Cell Staining Buffer
Quantity 250 mL
For Use With (Application) Flow Cytometry, Direct Immunofluorescence Staining, Indirect Immunofluorescence Staining, Intracellular Staining, Immunocytochemistry (ICC)
Form Unlabeled Solution
Content And Storage 2° to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.