Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Fc Mouse anti-Human, Rhesus Ma, Chimpanzee, Unconjugated, Clone: JDC-10, Southern Biotech™
SDP

Catalog No. OB904001 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB9040-01 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB9040-01 Supplier Southern Biotechnology Associates Supplier No. 904001
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SouthernBiotech monoclonal secondary antibodies offer high sensitivity and low background for use in a variety of applications.

Specifications

Antigen IgG Fc
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoassay, Western Blot
Classification Monoclonal
Clone JDC-10
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
Host Species Mouse
Immunogen Purified human IgG1 myeloma protein
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Chimpanzee, Human, Rhesus Monkey
Content And Storage Store at 2-8°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.