Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Genetically Distinct Collagens: Bovine Type II
SDP

Catalog No. p-4907992
Click to view available options
Quantity:
0.25 mg
0.5 mg

Highly purified fibrous protein for a variety of applications

Bovine origin, Type II.

  • Can be used as a collagen standard, antigen for antibody production, and coating material in cell culture studies, and in the formation of collagen gels
  • Prepared via controlled and limited pepsin digestion; greater than 90% pure
  • Supplied as a 0.5mg/mL solution in 500mmol/L acetic acid or lyophilized

Specifications

For Use With (Application) Cell Culture Coating Material, Antigen for Antibody Production, Collagen Gel Formation, Collagen Standard
Purification Method Controlled and limited pepsin digestion, followed by selective salt precipitation
Quantity 0.25 mg
Source Bovine
Product Type Collagen, Bovine Type II
Content And Storage Store at 2°C to 8°C or in small aliquots at -20°C
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.