Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SouthernBiotech™ Genetically Distinct Collagens: Human Type III

Click to view available options
Quantity:
0.5 mg
Description
Human origin, Type III.
- Can be used as a collagen standard, antigen for antibody production, and coating material in cell culture studies, and in the formation of collagen gels
- Prepared via controlled and limited pepsin digestion; greater than 90% pure
- Supplied as a 0.5mg/mL solution in 500mmol/L acetic acid
Specifications
Specifications
Concentration | 0.5 mg/mL |
For Use With (Application) | Cell Culture Coating Material, Antigen for Antibody Production, Collagen Gel Formation, Collagen Standard |
Purification Method | Controlled and limited pepsin digestion, followed by selective salt precipitation |
Quantity | 0.5 mg |
Source | Human |
Product Type | Collagen, Human Type III |
Content And Storage | Store at 2°C to 8°C or in small aliquots at -20°C |
Purity or Quality Grade | >90% by SDS-PAGE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction