Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Neutralite Avidin
SDP

Catalog No. p-3343042 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
1 mg
5 mg
1 mL
Conjugate:
AP
APC
Cy3
Cy5
FITC (Fluorescein)
HRP
R-phycoerythrin
Rhodamine
Texas Red
Unconjugated
beta-Galactosidase
pH:
7.4
8
8.1
Color:
Blue
Blue to Violet
Clear
Colorless to Light Brown
Light to Dark Amber
Light to Dark Pink
Light to Dark Yellow
Red to Dark Violet
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate pH Color
OB7200-02 1 mg FITC (Fluorescein) 7.4 Light to Dark Yellow
OB7200-04 1 mL AP 8 Colorless to Light Brown
OB7200-01 5 mg Unconjugated 8.1 Clear
OB7200-03 1 mg Rhodamine 7.4 Red to Dark Violet
OB720006 1 mL beta-Galactosidase 7.4 Colorless to Light Brown
OB720007 1 mg Texas Red 7.4 Blue to Violet
OB7200-11 0.1 mg APC 7.4 Blue
OB720015 1 mg Cy5 7.4 Blue
OB7200-05 1 mL HRP 7.4 Light to Dark Amber
OB7200-09 0.5 mg R-phycoerythrin 7.4 Light to Dark Pink
OB720012 0.1 mg Cy3 7.4 Light to Dark Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Reagents contain sodium azide. Sodium azide is very toxic if ingested or inhaled. Avoid contact with skin, eyes, or clothing. Wear eye or face protection when handling. If skin or eye contact occurs, wash with copious amounts of water. If ingested or inhaled, contact a physician immediately. Sodium azide yields toxic hydrazoic acid under acidic conditions. Dilute azide-containing compounds in running water before discarding to avoid accumulation of potentially explosive deposits in lead or copper plumbing.
Catalog No. OB7200-02 Supplier SouthernBiotech™ Supplier No. 720002
Only null left
Add to Cart
Add to Cart
Reagents contain sodium azide. Sodium azide is very toxic if ingested or inhaled. Avoid contact with skin, eyes, or clothing. Wear eye or face protection when handling. If skin or eye contact occurs, wash with copious amounts of water. If ingested or inhaled, contact a physician immediately. Sodium azide yields toxic hydrazoic acid under acidic conditions. Dilute azide-containing compounds in running water before discarding to avoid accumulation of potentially explosive deposits in lead or copper plumbing.

Neutralized, deglycosylated avidin purified from egg white

For Research Use Only. Not for Diagnostic or Therapeutic Use.

Flow cytometry; Enzyme-Linked-Immunosorbent-Assay (ELISA); Immunoblotting; Immunohistochemistry

Order Info

To ensure lot-to-lot consistency, each batch of product is tested by flow cytometry, particle concentration, FLISA or ELISA to conform to characteristics of a standard reference reagent.

Specifications

Color Light to Dark Yellow
Concentration 1 mg/mL
Conjugate FITC (Fluorescein)
pH 7.4
Quantity 1 mg
Source Egg White
Content And Storage 2°C to 8°C
Form Liquid
For Use With (Application) ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry
Specificity d-Biotin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.