Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Rat anti-Human, Rhesus Ma, Cynomolgus, Biotin, Clone: BVD2-21C11, Southern Biotech™
SDP

Catalog No. OB1011208 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB10112-08 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB10112-08 Supplier Southern Biotechnology Associates Supplier No. 1011208
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SouthernBiotech cytokine antibodies offer high sensitivity and low background for use in a variety of applications.

Specifications

Antigen GM-CSF
Applications ELISA, Dot Blot, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunoassay
Classification Monoclonal
Clone BVD2-21C11
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Supplied in PBS/NaN3
Host Species Rat
Immunogen E. coli-expressed human GM-CSF
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Cynomolgus, Human, Rhesus Monkey
Content And Storage Store at 2-8°C
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.