Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOWAHA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564177
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564177 Supplier Invitrogen™ Supplier No. PA564177
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRRLLGDPGL RGTTEPDATG GGSGSLAARR PVQVAATILS STTSAFLGVL ADDLMLQD Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SOWAHA
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SOWAHA
Gene Accession No. Q2M3V2
Gene Alias ANKRD43; ankyrin repeat domain-containing protein 43; ankyrin repeat domain-containing protein SOWAHA; protein sosondowah homolog A; sosondowah ankyrin repeat domain family member A; SOWAHA
Gene Symbols SOWAHA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SOWAHA. Recombinant protein control fragment (Product #RP-103566).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134548
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.