Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX-11 Monoclonal Antibody (SOX11-C1), Abbomax

Catalog No. 502204430
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4430 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4430 Supplier Abbomax Inc Supplier No. 606260
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Fetal brain tissue lysate Cellular location: Nucleus.

SOX-11 Monoclonal antibody specifically detects SOX-11 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SOX-11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone SOX11-C1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene SOX11
Gene Accession No. P35716
Gene Alias 1110038H03Rik; 6230403H02Rik; AI836553; end1; MRD27; Sox11; Sox-11; SRY; SRY (sex determining region Y)-box 11; SRY (sex-determining region Y)-box 11; SRY-box 11; SRY-box containing gene 11; SRY-box transcription factor 11; SRY-related HMG-box gene 11; transcription factor SOX-11
Host Species Mouse
Immunogen A recombinant protein
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6664
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.