Learn More
Description
Specifications
Specifications
| Antigen | Sox-11 |
| Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 19B2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. |
| Host Species | Rat |
| Immunogen | GST/His-tagged recombinant fragment corresponding to 113 amino acids from the C-terminal half of mouse Sox-11. |
| Purification Method | Protein G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.