Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sox-9 Rabbit anti-Human, Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE286825UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE2868-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE2868-25UL Supplier MilliporeSigma Supplier No. ABE286825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Sox-9 is a highly specific rabbit polyclonal antibody, that targets Transcription factor SOX-9 and has been tested in Immunofluorescence, Immunocytochemistry, Western Blotting and Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sox-9
Applications Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols SOX9
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 24 amino acids from the C-terminal end of human Sox-9.
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit ployclonal antibody detects SOX-9 in human, mouse, and rat cells. It targets an epitope with in 24 amino acids from the C-terminal end.
Target Species Human, Rat, Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.