Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224486H
Description
SOX1 Polyclonal specifically detects SOX1 in Human, Mouse, Equine, Opossum, Primate, Xenopus samples. It is validated for Western Blot.Specifications
SOX1 | |
Polyclonal | |
Western Blot | |
SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS with No Preservative | |
SOX1 | |
A portion of amino acids 100-150 of human SOX1 was used as the immunogen. | |
0.1 mL | |
Stem Cells | |
Human, Mouse, Equine, Opossum, Primate, Xenopus species | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction