Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $438.00

Specifications

Antigen SOX1
Concentration 0.5 mg/mL
Dilution Western Blot reported by customer review
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224486SS
SDP
View Documents
Novus Biologicals
NBP224486SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224486
SDP
View Documents
Novus Biologicals
NBP224486
0.1 mg
Each for $438.00
Only null left
Add to Cart
 
Description

Description

SOX1 Polyclonal specifically detects SOX1 in Human, Mouse, Equine, Opossum, Primate, Xenopus samples. It is validated for Western Blot.
Specifications

Specifications

SOX1
Western Blot reported by customer review
Polyclonal
Rabbit
Stem Cells
PBS, 0.05% BSA with 0.05% Sodium Azide
SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1
SOX1
IgG
Affinity Purified
The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, macaque, orangutan, gorilla, chimpanzee, cow, and opossum, 95% homologous in dog and wild boar, 90% homologous in cat and rat, and 85% homologous in mouse.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Equine, Opossum, Primate, Xenopus species
Q80XF2
6656
A portion of amino acids 100-150 of human SOX1 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.