Learn More
Invitrogen™ SOX1 Polyclonal Antibody

Description
Predicted to react with Bovine, Opossum, Pufferfish, Xenopus, Chimpanzee (100%).
Specifications
Specifications
| Antigen | SOX1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | SOX1 |
| Gene Accession No. | O00570, P53783 |
| Gene Alias | BB176347; LOC100912302; sex determining region Y-box 1; Sox1; Sox-1; sox1.S; SRY (sex determining region Y)-box 1; SRY box 1; SRY-box 1; SRY-box 1 S homeolog; SRY-box containing gene 1; SRY-box containing protein; SRY-box transcription factor 1; SRY-related HMG-box gene 1; transcription factor SOX-1; transcription factor SOX-1-like; XELAEV_18015596mg; XlSox1; xSox1 |
| Gene Symbols | SOX1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.