Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX1 Recombinant Rabbit Monoclonal Antibody (9L8Z4)
GREENER_CHOICE

Catalog No. PIMA538150
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538150 Supplier Invitrogen™ Supplier No. MA538150
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: A-549 Immunogen sequence: MYSMMMETDL HSPGGAQAPT NLSGPAGAGG GGGGGGGGGG GGGAKANQDR VKRPMNAFMV WSRGQRRKMA QENPKMHNSE ISKRLGAEWK VMSEAEKRPF.

SOX1, also known as SRY (sex determining region Y)-box 1, is a transcription factor belonging to the SRY-related HMG box family of transcription factors with a HMG box DNA-binding domain. SOX1 works through multiple independent pathways to promote neuronal cell fate determination and differentiation. This nuclear protein acts as a modulator of LINE retroposons promoter activity and is also involved in the development of the central nervous system, mainly neural and lens development. Transcriptional activator SOX1 binds directly to the HES1 promoter and suppresses HES1 transcription, thus attenuating Notch signaling. It also binds to beta-catenin and suppresses beta-catenin-mediated TCF/LEF signaling, thus potentially attenuating the WNT signaling pathway. Expression of SOX1 is seen mostly in embryonic tissues and moderately in brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9L8Z4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SOX1
Gene Accession No. O00570
Gene Alias BB176347; LOC100912302; sex determining region Y-box 1; Sox1; Sox-1; sox1.S; SRY (sex determining region Y)-box 1; SRY box 1; SRY-box 1; SRY-box 1 S homeolog; SRY-box containing gene 1; SRY-box containing protein; SRY-box transcription factor 1; SRY-related HMG-box gene 1; transcription factor SOX-1; transcription factor SOX-1-like; XELAEV_18015596mg; XlSox1; xSox1
Gene Symbols SOX1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SOX1 (O00570).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100912302, 6656
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.