Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318510
This item is not returnable.
View return policy
Description
SOX10 Polyclonal antibody specifically detects SOX10 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
SOX10 | |
Polyclonal | |
Unconjugated | |
Tris-citrate/phosphate pH 7 -8. | |
Rabbit | |
Affinity purified | |
RUO | |
6663 | |
Store at 4°C. Do not freeze. | |
IgG |
Western Blot, Immunoprecipitation | |
1 mg/ml | |
Western Blot 1:10000 - 1:25000, Immunoprecipitation 50-100 ug/mg lysate | |
MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of | |
Recognizes region between residues Between between 416 and 466.. | |
100 μg | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction