Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX10 Mouse anti-Human, Mouse, Clone: SOX10/991, Abnova™

Catalog No. 89034525 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-525 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-525 Supplier Abnova Corporation Supplier No. MAB14442
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human SOX10.

  • High specificity
  • High reactivity

Specifications

Antigen SOX10
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SOX10/991
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human SOX10.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end u
Formulation In 10 mM PBS (0.05% BSA and 0.05% azide).
Gene Accession No. P56693
Gene Alias DOM/MGC15649/WS2E/WS4
Gene Symbols SOX10
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 115-269 of human SOX10.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6663
Test Specificity Recognizes a protein of ∼55kDa, identified as SOX10. This antibody is highly specific and does not cross-react with other members of the SOX-family.
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.