Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX10 Recombinant Rabbit Monoclonal Antibody (SD204-04)
GREENER_CHOICE

Catalog No. PIMA532398
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532398 Supplier Invitrogen™ Supplier No. MA532398
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

SOX10 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. This protein may act as a transcriptional activator after forming a protein complex with other proteins. It acts as a nucleocytoplasmic shuttle protein and is important for neural crest and peripheral nervous system development. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Western Blot
Classification Recombinant Monoclonal
Clone SD204-04
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene SOX10
Gene Accession No. O55170, P56693, Q04888
Gene Alias cls; cls/sox10; colorless; colourless; colourless/sox10; DOM; dominant megacolon; dominant megacolon, mouse, human homolog of; golas; gos; HMG-box transcription factor Sox10; MGC15649; PCWH; Protein SOX-21; SOX10; Sox-10; sox10 protein; sox10b; Sox21; SOX9; SRY; SRY (sex determining region Y)-box 10; SRY (sex determining region Y)-box 9 (campomelic dysplasia, autosomal sex-reversal); SRY box 10; SRY-box 10; SRY-box containing gene 10; SRY-box containing protein 10; SRY-box transcription factor 10; SRY-related HMG-box gene 10; transcription factor SOX-10; Transcription factor SOX-M; WS2E; WS4; WS4C; zgc:100757
Gene Symbols SOX10
Host Species Rabbit
Immunogen Synthetic peptide Human SOX10 aa 411-460.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20665, 29361, 6663
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.