Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX11 Antibody (SOX11/3235R), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308594AF750
Description
SOX11 Monoclonal antibody specifically detects SOX11 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SOX11 | |
Monoclonal | |
Alexa Fluor 750 | |
SRY (sex determining region Y)-box 11, SRY (sex-determining region Y)-box 11, SRY-related HMG-box gene 11, transcription factor SOX-11 | |
Recombinant fragment (around aa300-400) of human SOX11 protein (Uniprot: P35716 ) | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
SOX11/3235R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
6664 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction