Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SOX11 Recombinant Rabbit Monoclonal Antibody (5K1J5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551319
Description
Immunogen sequence: MVQQAESLEA ESNLPREALD TEEGEFMACS PVALDESDPD WCKTASGHIK RPMNAFMVWS KIERRKIMEQ SPDMHNAEIS KRLGKRWKML KDSEKIPFIR.
This intronless gene encodes a member of the SOX family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may function in the developing nervous system and play a role in tumorigenesis.
Specifications
SOX11 | |
Recombinant Monoclonal | |
0.51 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P0C1G9, P35716, Q7M6Y2 | |
SOX11 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SOX11 (NP_003099.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
Liquid |
ELISA, Western Blot | |
5K1J5 | |
Unconjugated | |
SOX11 | |
1110038H03Rik; 6230403H02Rik; AI836553; end1; MRD27; Sox11; Sox-11; SRY; SRY (sex determining region Y)-box 11; SRY (sex-determining region Y)-box 11; SRY-box 11; SRY-box containing gene 11; SRY-box transcription factor 11; SRY-related HMG-box gene 11; transcription factor SOX-11 | |
Rabbit | |
Affinity chromatography | |
RUO | |
20666, 6664, 84046 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction