Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SOX12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566407
Description
Immunogen sequence: DEELLEVRLVE TPGRELWRMV PAGRAARGQA E Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
Members of the SOX family of transcription factors are characterized by the presence of a DNA-binding high mobility group (HMG) domain, homologous to the HMG box of sex-determining region Y (SRY). Forming a subgroup of the HMG domain superfamily, SOX proteins have been implicated in cell fate decisions in a diverse range of developmental processes. SOX transcription factors have diverse tissue-specific expression patterns during early development and have been proposed to act as target-specific transcription factors and/or as chromatin structure regulatory elements. The protein encoded by this gene was identified as a SOX family member based on conserved domains, and its expression in various tissues suggests a role in both differentiation and maintenance of several cell types.
Specifications
SOX12 | |
Polyclonal | |
Unconjugated | |
SOX12 | |
2010205A02Rik; Protein SOX-22; SOX12; Sox-12; SOX22; SOX-22 protein; SRY (sex determining region Y)-box 12; SRY box 12; SRY-box 12; SRY-box containing gene 12; SRY-related HMG-box gene 22; transcription factor SOX-12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6666 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15370 | |
SOX12 | |
Recombinant Human SOX12. Recombinant protein control fragment (Product #RP-107083). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction