Learn More
Invitrogen™ SOX13 Polyclonal Antibody

Description
Immunogen sequence: DVLYPRAAGM PLAQPLVEHY VPRSLDPNMP VIVNTCSLRE EGEGTDDRHS VADGEMYRYS EDEDSEGEEK SDGELVVLT Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 87%.
Specifications
Specifications
| Antigen | SOX13 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SOX13 |
| Gene Accession No. | Q9UN79 |
| Gene Alias | AA407916; AW259412; AW540933; ICA12; islet cell antibody 12; islet cell antigen 12; MGC117216; mSox13; RP11-74C13.1; SOX13; Sox-13; SRY (Sex determining region Y)-box 13; SRY box 13; SRY-box 13; SRY-box containing gene 13; SRY-related HMG-box gene 13; transcription factor SOX-13; Type 1 diabetes autoantigen ICA12 |
| Gene Symbols | SOX13 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.