Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111403 Supplier Invitrogen™ Supplier No. PA5111403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDARQLLHSH SGALDGSPNT PFRKDLISLD SSPAKERLED GCVHPLEEAM LSCDMDGSRH FPESRNSSHI Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 93%.

This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. It has also been determined to be a type-1 diabetes autoantigen, also known as islet cell antibody 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX13
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SOX13
Gene Accession No. Q9UN79
Gene Alias AA407916; AW259412; AW540933; ICA12; islet cell antibody 12; islet cell antigen 12; MGC117216; mSox13; RP11-74C13.1; SOX13; Sox-13; SRY (Sex determining region Y)-box 13; SRY box 13; SRY-box 13; SRY-box containing gene 13; SRY-related HMG-box gene 13; transcription factor SOX-13; Type 1 diabetes autoantigen ICA12
Gene Symbols SOX13
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human SOX13. Recombinant protein control fragment (Product #RP-106579).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9580
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.