Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SOX13 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | SOX13 |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111341
|
Invitrogen™
PA5111341 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111403
|
Invitrogen™
PA5111403 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: DVLYPRAAGM PLAQPLVEHY VPRSLDPNMP VIVNTCSLRE EGEGTDDRHS VADGEMYRYS EDEDSEGEEK SDGELVVLT.
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. It has also been determined to be a type-1 diabetes autoantigen, also known as islet cell antibody 12.Specifications
SOX13 | |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UN79 | |
9580 | |
Recombinant Protein corresonding to Human SOX13. Recombinant protein control fragment (Product #RP-104934). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
SOX13 | |
AA407916; AW259412; AW540933; ICA12; islet cell antibody 12; islet cell antigen 12; MGC117216; mSox13; RP11-74C13.1; SOX13; Sox-13; SRY (Sex determining region Y)-box 13; SRY box 13; SRY-box 13; SRY-box containing gene 13; SRY-related HMG-box gene 13; transcription factor SOX-13; Type 1 diabetes autoantigen ICA12 | |
SOX13 | |
Primary | |
Antigen affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title