Learn More
Invitrogen™ SOX14 Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Goat: 100%; Guinea Pig: 86%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%; Zebrafish: 79%.
Specifications
Specifications
| Antigen | SOX14 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | SOX14 |
| Gene Accession No. | O95416 |
| Gene Alias | HMG box transcription factor SOX-14; Protein SOX-28; SOX14; Sox-14; SOX28; SRY (sex determining region Y)-box 14; SRY-box 14; SRY-box containing gene 14; transcription factor SOX-14 |
| Gene Symbols | SOX14 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.