Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SOX14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598168
Description
SOX14 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
This intronless gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. Mutations in this gene are suggested to be responsible for the limb defects associated with blepharophimosis, ptosis, epicanthus inversus syndrome (BPES) and Mobius syndrome.
Specifications
SOX14 | |
Polyclonal | |
Unconjugated | |
SOX14 | |
HMG box transcription factor SOX-14; Protein SOX-28; SOX14; Sox-14; SOX28; SRY (sex determining region Y)-box 14; SRY-box 14; SRY-box containing gene 14; transcription factor SOX-14 | |
Rabbit | |
Protein G | |
RUO | |
8403 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O95416 | |
SOX14 | |
Recombinant Human Transcription factor SOX-14 protein (1-137AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction