Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110413
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110413 Supplier Invitrogen™ Supplier No. PA5110413
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RYVFPLPYLG DTDPLKAAGL PVGASDGLLS APEKARAFLP PASAPYSLLD PAQFSSSAIQ KMGEVPHTLA TGALPYASTL GYQNGAFGSL SCPSQHTHTH PSPTNPGYVV PCNCTAWSAS TLQPPVAYIL FPGMTKTGID PYSSAHATAM.

This intronless gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. Mutations in this gene are suggested to be responsible for the limb defects associated with blepharophimosis, ptosis, epicanthus inversus syndrome (BPES) and Mobius syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX14
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SOX14
Gene Accession No. Q04892
Gene Alias HMG box transcription factor SOX-14; Protein SOX-28; SOX14; Sox-14; SOX28; SRY (sex determining region Y)-box 14; SRY-box 14; SRY-box containing gene 14; transcription factor SOX-14
Gene Symbols SOX14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 91-240 of human SOX14 (NP_004180.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20669, 300954
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.