Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SOX15 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518846
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
SOX15 is a member of the SOX (SRY-related HMG-box) family of transcription factors and is involved in the regulation of embryonic development and in the determination of cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins.
Specifications
| SOX15 | |
| Polyclonal | |
| Unconjugated | |
| SOX15 | |
| Protein SOX-12; Protein SOX-15; Protein SOX-20; SOX12; SOX15; Sox-15; SOX20; SOX26; SOX27; SRY (sex determining region Y)-box 15; SRY (sex determining region Y)-box 20; SRY box 15; SRY-box 15; SRY-box containing gene 15; SRY-box transcription factor 15 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 6665 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O60248 | |
| SOX15 | |
| Synthetic peptide sequence (CSLPQSDPRLQGE) corresponding to the internal amino acids of SOX15 (aa 190-203). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction