Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541519 Supplier Thermo Scientific Supplier No. PA541519
Add to Cart
Add to Cart

SOX15 Polyclonal Antibody

SOX15 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX15
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SOX15
Gene Accession No. O60248
Gene Alias Protein SOX-12; Protein SOX-15; Protein SOX-20; SOX12; SOX15; Sox-15; SOX20; SOX26; SOX27; SRY (sex determining region Y)-box 15; SRY (sex determining region Y)-box 20; SRY box 15; SRY-box 15; SRY-box containing gene 15; SRY-box transcription factor 15
Gene Symbols SOX15
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SOX15.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6665
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.