Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548044
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548044 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548044 Supplier Invitrogen™ Supplier No. PA548044
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In Western blots, less than 1% cross-reactivity with recombinant human (rh) SOX1, rhSOX2, rhSOX5, rhSOX6, rhSOX7, rhSOX10, rhSOX12, rhSOX14, rhSOX15, rhSOX17, and rhSOX21 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

SOX15 is a member of the SOX (SRY-related HMG-box) family of transcription factors and is involved in the regulation of embryonic development and in the determination of cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX15
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene SOX15
Gene Accession No. O60248
Gene Alias Protein SOX-12; Protein SOX-15; Protein SOX-20; SOX12; SOX15; Sox-15; SOX20; SOX26; SOX27; SRY (sex determining region Y)-box 15; SRY (sex determining region Y)-box 20; SRY box 15; SRY-box 15; SRY-box containing gene 15; SRY-box transcription factor 15
Gene Symbols SOX15
Host Species Sheep
Immunogen E. coli-derived recombinant human SOX15 Met1-Leu233.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6665
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.