Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX17 Antibody (614013), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB19241F
Description
SOX17 Monoclonal antibody specifically detects SOX17 in Human samples. It is validated for ImmunocytochemistrySpecifications
SOX17 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
64321 | |
Store at 4°C in the dark. | |
IgG1 |
Immunocytochemistry | |
614013 | |
Immunocytochemistry | |
FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3 | |
E. coli-derived recombinant human SOX17, Asp177-Val414, Accession # Q9H6I2 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction