Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX17 Mouse anti-Human, Biotin, Clone: OTI5G6, liquid, TrueMAB™
SDP

Catalog No. 501677662
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7662 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7662 Supplier OriGene Supplier No. TA700126
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SOX17, also known as transcription factor SOX-17, as the name suggests is a transcription factor or a probable transcriptional activator in the premeiotic germ cells. It belongs to the SRY-related HMG box family of transcription factors with a high-mobility group DNA-binding domain (HMG box), that plays a key role in embryogenesis and binds to the sequences 5′-AACAAT-′3 or 5′-AACAAAG-3′. SOX17 plays an essential role in cardiac mesoderm specification as well as in definitive hematopoiesis, maintaining fetal hematopoietic stem cells, hematopoiesis in the fetal liver and yolk sac. Along with beta-catenin, it is known to cooperate in the regulation of transcription of endodermal genes. Defects in SOX17are known to cause a severe defects in fetal hematopoiesis.

Specifications

Antigen SOX17
Applications ELISA
Classification Monoclonal
Clone OTI5G6
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 10mg/ml BSA, 50% glycerol and 0.05% sodium azide
Gene SOX17
Gene Accession No. Q9H6I2
Gene Alias VUR3
Gene Symbols SOX17
Host Species Mouse
Immunogen Recombit protein expressed in E.coli corresponding to amino acids 177-414 of human SOX17
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64321
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.