Learn More
Sox17 Mouse anti-Human, Clone: OTI5G6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500282

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SOX17, also known as transcription factor SOX-17, as the name suggests is a transcription factor or a probable transcriptional activator in the premeiotic germ cells. It belongs to the SRY-related HMG box family of transcription factors with a high-mobility group DNA-binding domain (HMG box), that plays a key role in embryogenesis and binds to the sequences 5′-AACAAT-′3 or 5′-AACAAAG-3′. SOX17 plays an essential role in cardiac mesoderm specification as well as in definitive hematopoiesis, maintaining fetal hematopoietic stem cells, hematopoiesis in the fetal liver and yolk sac. Along with beta-catenin, it is known to cooperate in the regulation of transcription of endodermal genes. Defects in SOX17are known to cause a severe defects in fetal hematopoiesis.Specifications
Sox17 | |
Monoclonal | |
Unconjugated | |
SOX17 | |
VUR3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
64321 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5G6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H6I2 | |
SOX17 | |
Recombit protein expressed in E.coli corresponding to amino acids 177-414 of human SOX17 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.