Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sox17, Mouse Anti-Human, Oligo, Clone: P7-969, BD AbSeq™
Mouse Oligoclonal Antibody
Supplier: BD Biosciences 940511
Description
Sox17 Oligoclonal antibody specifically detects Sox17 in Human (Tested in Development) samples. It is validated for Single Cell 3'Sequencing
Specifications
Sox17 | |
Oligoclonal | |
Unconjugated | |
SOX-17, SOX17, FLJ22252 | |
Human Sox17 Recombinant Protein | |
RUO | |
64321 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Single Cell 3' Sequencing | |
P7-969 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction