Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572815
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572815 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572815 Supplier Invitrogen™ Supplier No. PA572815
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SOX17 Polyclonal Antibody for ELISA, IHC, Western Blot

SOX17 (transcription factor SOX-17) belongs to the SRY-related HMB box family of transcription factors. It acts as a transcription regulator that binds target promoter DNA and bends the DNA. It binds to the sequences 5'-AACAAT-'3 or 5'-AACAAAG-3'. SOX17 modulates transcriptional regulation via WNT3A and inhibits Wnt signaling. It promotes degradation of activated CTNNB1. SOX17 plays an essential role in the regulation of embryonic development and is required for normal looping of the embryonic hear tube and for normal development of the definitive gut endoderm. Mutations in the gene can result in vesicoureteral reflux.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX17
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SOX17
Gene Accession No. Q61473, Q9H6I2
Gene Alias HMG transcription factor SOX17; HMG-box transcription factor Sox17; Sox17; Sox-17; SRY (sex determining region Y)-box 17; SRY box 17; SRY-box 17; SRY-box containing gene 17; SRY-box transcription factor 17; SRY-related HMG-box transcription factor SOX17; transcription factor SOX-17; transcription factor SOX-17; LOW QUALITY PROTEIN: transcription factor SOX-17; VUR3
Gene Symbols SOX17
Host Species Rabbit
Immunogen SOX17 antibody for a 16 amino acid peptide near the center of human SOX17. The immunogen is within amino acids 240 - 290 of SOX17.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20671, 312936, 64321
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.