Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX17 Polyclonal Antibody, Huabio

Catalog No. 502544534
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4534 Supplier HUABIO RESEARCH INC Supplier No. HA500495
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SOX17 Polyclonal antibody specifically detects SOX17 in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SOX17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene SOX17
Gene Accession No. Q9H6I2
Gene Alias HMG transcription factor SOX17; HMG-box transcription factor Sox17; Sox17; Sox-17; SRY (sex determining region Y)-box 17; SRY box 17; SRY-box 17; SRY-box containing gene 17; SRY-box transcription factor 17; SRY-related HMG-box transcription factor SOX17; transcription factor SOX-17; transcription factor SOX-17; LOW QUALITY PROTEIN: transcription factor SOX-17; VUR3
Host Species Rabbit
Immunogen Synthetic peptide within human SOX17 aa 365-414.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 312936, 64321
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.