Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX17 Rabbit anti-Mouse, Rat, Human, Polyclonal, MilliporeSigma™

Catalog No. 09038IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-038-IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-038-IMI Supplier MilliporeSigma Supplier No. 09038I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-SOX17 is a highly specific rabbit polyclonal antibody that targets Transcription factor SOX-17 and has been tested in Immunohistochemistry (Paraffin), Immunofluorescence, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX17
Applications Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Sox-17
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal region of murine Sox17.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_035571
Test Specificity This polyclonal antibody detects Sox17 in human, mouse, and rat. It targets an epitope within 17 amino acids from the N-terminal region.
Target Species Mouse, Rat, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.