Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX18 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241289PCP
Description
SOX18 Polyclonal antibody specifically detects SOX18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
| SOX18 | |
| Polyclonal | |
| Western Blot, ELISA | |
| HLTS, SRY (sex determining region Y)-box 18, SRY-box 18, transcription factor SOX-18 | |
| Antibody was raised against a 17 amino acid peptide near the amino terminus of human SOX18. The immunogen is located within amino acids 40 - 90 of SOX18. Amino Acid Squence: ASPPSPQRSPPRSPEPG | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 54345 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction