Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX18 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | SOX18 |
---|---|
Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SOX18 Polyclonal specifically detects SOX18 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
SOX18 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
HLTS, SRY (sex determining region Y)-box 18, SRY-box 18, transcription factor SOX-18 | |
The immunogen is a synthetic peptide directed towards the middle region of human SOX18 (NP_060889). Peptide sequence RTAPPAAPLAGLYYGTLGTPGPYPGPLSPPPEAPPLESAEPLGPAADLWA | |
Affinity purified |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
54345 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title