Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX18 Rabbit anti-Human, Unconjugated, Clone: DCD-19, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20736
Description
SOX18 Monoclonal antibody specifically detects SOX18 in Human samples. It is validated for WB, IHC.Specifications
SOX18 | |
Monoclonal | |
Unconjugated | |
SOX18 | |
Rabbit | |
100 μg | |
54345 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Western Blot, Immunohistochemistry | |
DCD-19 | |
SRY (sex determining region Y)-box 18 | |
HLTS | |
A synthetic peptide corresponding to human SOX18 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction