Learn More
SOX2 Antibody (4G8), Alexa Fluor™ 532, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP229623AF532
Catalog No. NP229623A32
Only null left
Add to Cart
Description
Description
SOX2 Monoclonal specifically detects SOX2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
SOX2
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
SOX2
Full length recombinant human SOX2 (NCBI NP_003097).
Primary
Human
IgG2b
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
4G8
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
Mouse
0.1 mL
6657
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.