Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX2 Antibody (4G8), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP229623C
Description
SOX2 Monoclonal specifically detects SOX2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SOX2 | |
| Monoclonal | |
| DyLight 650 | |
| ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human. Based on sequence homology, this antibody is predicted to react with Mouse (96%), Chicken (93%), Sheep (99%) and Monkey/Primate species (99%). Sequence homology with other species: Zebrafish (87%), Xenopus tropicalis (87%), Xenopus laevis (88%). | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4G8 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| SOX2 | |
| Full length recombinant human SOX2 (NCBI NP_003097). | |
| 0.1 mL | |
| Cellular Markers, Core ESC Like Genes, Embryonic Stem Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| 6657 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction