Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
Specifications
| Antigen | SOX2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry Assay dependent, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence Assay dependent, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11079875
![]() |
Novus Biologicals
NB11079875 |
0.1 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
SOX2 Polyclonal specifically detects SOX2 in Human, Mouse, Rat, Fish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SOX2 | |
| Western Blot 1:100-1:2000, Flow Cytometry Assay dependent, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence Assay dependent, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Rabbit | |
| Core ESC Like Genes, Embryonic Stem Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
| PBS with 0.02% Sodium Azide | |
| ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 | |
| SOX2 | |
| IgG | |
| Affinity Purified | |
| SOX-2 |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Fish | |
| P48431 | |
| 6657 | |
| Reacts with residues 113-127 of the 37 kDa (predicted molecular weight is 34 kDa) human, mouse and rat SOX-2 protein. Sequence is 100% conserved in most species. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 34.3 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title