Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX2 Antibody (SOX2/1791), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254594DL594
Description
SOX2 Monoclonal specifically detects SOX2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SOX2 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SOX2 | |
Recombinant fragment (within aa176-305) of human SOX2 protein (exact sequence is proprietary) (Uniprot: P48431) | |
0.1 mL | |
Cellular Markers, Core ESC Like Genes, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
Human | |
IgG2b κ |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SOX2/1791 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction