Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sox2, Mouse Anti-Human, Oligo, Clone: O30-678, BD AbSeq™
Mouse Oligoclonal Antibody
Supplier: BD Biosciences 940510
Description
Sox2 Oligoclonal antibody specifically detects Sox2 in Human,Mouse (Tested in Development) samples. It is validated for Single Cell 3'Sequencing
Specifications
| Sox2 | |
| Oligoclonal | |
| Unconjugated | |
| ANOP3; MCOPS3; SOX2; SRY (sex determining region Y)-box 2; SRY-related HMG-box gene 2; Sox2; transcription factor SOX-2 | |
| Human Sox2 Recombinant Protein | |
| RUO | |
| 6657, 20674 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Single Cell 3' Sequencing | |
| O30-678 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human, Mouse | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction