Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sox2 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$159.90 - $633.45
Specifications
| Antigen | Sox2 |
|---|---|
| Concentration | Conc. Not Determined |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-4121
![]()
|
Active Motif
39824 |
10 μL |
Each for $159.90
|
|
|||||
50-199-4120
![]()
|
Active Motif
39823 |
100 μL |
Each for $633.45
|
|
|||||
Description
SOX2 is an intronless gene encoding a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of the SOX2 gene is required for stem-cell maintenance in the central nervous system, and also regulates gene expression in the stomach. The SOX2 gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). Further, SOX2 protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in the SOX2 gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation, optic nerve hypoplasia and syndromic microphthalmia.Specifications
| Sox2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| P48432 | |
| 20674 | |
| This Sox2 antibody was raised against a peptide corresponding to amino acids amino acids 111-140 mouse Sox2. | |
| Primary | |
| Antibody |
| Conc. Not Determined | |
| Polyclonal | |
| Liquid | |
| RUO | |
| whole serum with 30% glycerol and 0.035% sodium azide; pH | |
| ANOP3; cb236; lcc; MCOPS3; RGD1565646; sex determining region Y-box 2; sox2; Sox-2; soxp; SRY (sex determining region Y)-box 2; SRY box 2; SRY-box 2; SRY-box containing gene 2; SRY-related HMG-box gene 2; transcription factor SOX2; Transcription factor SOX-2; wu:fb83g04; wu:fc14d07; ysb; zgc:65860; zgc:77389 | |
| SOX2 | |
| IgG | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| SOX2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title