Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX2 Rabbit anti-Human, Mouse, Non-human Primate, Porcine, Polyclonal, FabGennix

Catalog No. 502209200
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9200 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9200 Supplier Fabgennix Inc Supplier No. SOX2201AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SOX2 Polyclonal antibody specifically detects SOX2 in Human, Mouse, Non-human Primate, Porcine samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen SOX2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene SOX2
Gene Accession No. P48431, P48432
Gene Alias ANOP3; cb236; lcc; MCOPS3; RGD1565646; sex determining region Y-box 2; SOX 2; SOX2; Sox-2; soxp; SRY (sex determining region Y)-box 2; SRY box 2; SRY-box 2; SRY-box 2; SOX2; SRY-box containing gene 2; SRY-box transcription factor 2; SRY-related HMG-box gene 2; transcription factor SOX2; transcription factor SOX-2; wu:fb83g04; wu:fc14d07; ysb; zgc:65860; zgc:77389
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 50-100on human Sex determining region Y-box 2 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20674, 407739, 460869, 6657
Target Species Human, Mouse, Non-human Primate, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.