Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX2 Rabbit anti-Human, Mouse, Unconjugated, Clone: SOX2/3169R, Abnova™

Catalog No. 89339605 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-605 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-605 Supplier Abnova Corporation Supplier No. RAB00626
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

SOX2 Monoclonal antibody specifically detects SOX2 in Human, Mouse samples. It is validated for WB, ELISA, IHC-P.

Specifications

Antigen SOX2
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SOX2/3169R
Conjugate Unconjugated
Description SRY (sex determining region Y)-box 2
Gene SOX2
Gene Alias ANOP3, MCOPS3, MGC2413
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 176-305 of human SOX2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6657
Target Species Human, Mouse
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.