Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX21 Antibody (CL4688), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $646.00
Specifications
Antigen | SOX21 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
SOX21 Monoclonal specifically detects SOX21 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SOX21 | |
Monoclonal | |
Purified | |
RUO | |
SOX25SOX-A, SOXA, SRY (sex determining region Y)-box 21, SRY-box 21, transcription factor SOX-21 | |
SOX21 | |
IgG2a | |
Protein A purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Mouse | |
11166 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GKPQLDPYPAAYAAAL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title