Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX21 Monoclonal Antibody (CL4681)
GREENER_CHOICE

Catalog No. PIMA531402
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531402 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531402 Supplier Invitrogen™ Supplier No. MA531402
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: GKPQLDPYPA AYAAAL Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

The SOX gene family is defined by the Sry-related HMG box domain that mediates sequence-specific DNA binding. A variety of functions are associated with the SOX proteins during development. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of this family have been shown to be conserved during evolution and to play key roles during animal development. SOX21 may play a role as an activator of transcription of OPRM1. Overexpression of SOX21 can cause an up-regulation of the OPRM1 distal promoter in mor-expressing neuronal cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX21
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL4681
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sox21
Gene Accession No. Q811W0, Q9Y651
Gene Alias DNA-binding protein SOX21; HMG-box protein SOX21; Sox21; Sox25; SOX25SOX-A; SOXA; SOX-A; SRY (sex determining region Y)-box 21; SRY box 21; SRY-box 21; SRY-box containing gene 21; transcription factor SOX-21
Gene Symbols Sox21
Host Species Mouse
Immunogen Synthetic peptide corresponding to Human SOX21.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11166, 223227
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.